To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Ring Finger Protein 2
- Antigen Symbol:RNF2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Frog,Chicken,Orangutan,Chimpanzee,Human,Bovine,Dog,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:6045
- Antigen Synonyms:HIPI3|RING1B|BAP-1|RING2|DING|BAP1
- Amino Acid Number:189 to 201
- Storage Buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide,
- Storage Temperature:–20 °C
- Concentration:1.0 mg/ml by UV absorbance at 280 nm
- Shipping Temperature:Dry Ice
- Immunogen:RING1B Antibody was prepared from whole goat serum produced by repeated immunizations with a synthetic peptide corresponding aa 189-201 of human RING1B Protein.
- Purification:affinity-purified
- Cat. No.:RL600-101-292
- Supplier no.:600-101-292
Specifications
About this item
Anti-RING1B purified antibody has been tested for use in ELISA and by western blot. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 38 kDa in size corresponding to RING1B by western blotting.
Recommended Dilutions: ELISA: 1:5000 - 1:25000; Immunofluorescence: 1:300; Western Blot: 1:500 - 1:2000;
Type: Primary
Antigen: RNF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Bovine, Chicken, Chimpanzee, Dog, Human, Mouse, Rat, frog, orangutan