Anti-SPRY4 Rabbit Polyclonal Antibody
Catalog # RL600-401-697
Supplier: Rockland Immunochemical
Specifications
- Antibody Type:Primary
- Antigen Name:Sprouty RTK Signaling Antagonist 4
- Antigen Symbol:SPRY4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Chimpanzee,Human
- Western Blot:Yes
- Size:100 µg
- Cross Adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:81848
- Antigen Synonyms:Spry-4|Protein sprouty homolog 4|SPRY4
- Amino Acid Number:306 to 322
- Storage Buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide,
- Storage Temperature:–20 °C
- Concentration:1.1 mg/ml by UV absorbance at 280 nm
- Shipping Temperature:Dry Ice
- Immunogen:This affinity purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to amino acids 306-322 of Human Sprouty-4 Protein.
- Purification:affinity-purified
- Cat. No.:RL600-401-697
- Supplier no.:600-401-697
Specifications
About this item
This affinity purified antibody has been tested for use in ELISA, immunochemistry and western blot. Specific conditions for reactivity should be optimized by the end user.Expect a band (maybe a doublet) ~35 kDa in size.
Recommended Dilutions: ELISA: 1:3000 - 1:12000; Western Blot: 1:100 - 1:1000; Immunochemistry: 1 µg/ml to 10 µg/ml;
Type: Primary
Antigen: SPRY4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Chimpanzee, Human