Anti-CBLC Rabbit Polyclonal Antibody
100 µg
Catalog # RL600-401-889
Supplier: Rockland Immunochemical
Supplier #: 600-401-889
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
This affinity purified antibody has been tested for use in ELISA, immunochemistry, immunoprecipitation and western blot. Specific conditions for reactivity should be optimized by the end user. Expect a band at ~52 kDa in size.
Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:500 - 1:3000; Immunochemistry: 1:500 - 1:3000;
Type: Primary
Antigen: CBLC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Specifications
- Size:100 µg
- Clonality:Polyclonal
- ImmunoPrecipitation:Yes
- Form:Liquid (sterile filtered)
- Format:Antibody fraction
- Antibody Type:Primary
- Gene ID:23624
- Immunogen:This affinity-purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to amino acids 444-458 of Human Cbl-c.
- Amino Acid Number:444 to 458
- Storage Temperature:–20 °C
- Cross Adsorption:No
- Purification:affinity-purified
- Storage Buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide,
- Isotype:IgG
- Host:Rabbit
- Concentration:1.42 mg/ml by UV absorbance at 280 nm
- Cat. No.:RL600-401-889
- Reactivity:Human
- Western Blot:Yes
- Shipping Temperature:Dry Ice
- Antigen Symbol:CBLC
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen Synonyms:RNF57|CBL-SL|CBL-3
- Antigen Name:Cbl proto-oncogene C, E3 ubiquitin protein ligase
- ELISA:Yes
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



