Nalgene® Crimp Finish Serum Vials, PETG, Non-Sterile, Thermo Scientific
13500-118
- Cap included:No
- Cap Size:20 mm
- Capacity:10.00 ml
- Color:Clear
- Cat. No.:13500-118
- Diameter ×H:24×50 mm
- Supplier no.:322030-0010
Nalgene non-sterile serum vials are made for use with stoppers and caps (not included).
- Non-cytotoxic
- Non-pyrogenic
- Packaged in tray less shrink-wrap modules
These serum vials are lighter in weight than glass, and protect against leakage, breakage, and contamination.
Int. Ø: 13 mm