ContactUsLinkComponent
Ready-to-Use Phosphate and Tris Buffer Solutions, G-Biosciences
Ready-to-Use Phosphate and Tris Buffer Solutions, G-Biosciences
Catalog # 89167-714
Supplier:  G-Biosciences
CAS Number:  
Ready-to-Use Phosphate and Tris Buffer Solutions, G-Biosciences
Catalog # 89167-714
Supplier:  G-Biosciences
Supplier Number:  786-027
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    PBS, Phosphate Buffered Saline [10X]
  • Size:
    500 mL
  • Cat. No.:
    89167-714
  • Supplier no.:
    786-027
Specifications
About this item
G-Biosciences' Phosphate and Tris buffered saline solutions are both conveniently supplied in a 10X concentrated, ready-to-use format.
G-Biosciences' Tris Buffered Saline (TBS) contains one to five percent 100mM Tris HCl (CAS# 77-86-1) and less than ten percent 1.5M sodium chloride (CAS# 7647-14-5). The solution is at a pH of 7.5.
G-Biosciences' Phosphate BufferedSane (PBS) contains 1.5M sodium chloride (CAS# 7647-14-5), 30mM potassium chloride (CAS# 7447-40-7), 80mM sodium phosphate monobasic, and 20mM potassium phosphate dibasic. The solution is at a pH of 7.4.
G-Biosciences also offers convenient, easy-to-use, pre-mixed dry powder buffer packs that simply require adding water to generate for use. G-Biosciences’ JAW™ buffered saline packs are offered in the following formats: Phosphate buffered saline packs (available as either 1X or 10X concentrated), Carbonate-Bicarbonate buffered packs (1X concentrated), or Tris buffered saline packs (available as either 1X or 20X concentrated).
Ordering information: Accessories available