Specifications
- Description:Phosphatase Assay kit
- Size:1000 assays
- Cat. No.:95029-244
- Supplier no.:786-453
Specifications
About this item
G-Biosciences' Phosphatase Assay kit is designed to measure the activity of phosphatases in biological samples and to screen for agonists and inhibitors of phosphatases.
G-Biosciences' Phosphatase Assay kit uses para-nitrophenyl phosphate (pNPP), a chromogenic substrate for most phosphatases, including alkaline phosphatases, acid phosphatases, protein tyrosine phosphatases and serine/threonine phosphatases. The phosphatases remove the phosphate group to generate p-nitrophenol, which is deprotonated under alkaline conditions in order to produce p-nitrophenolate, which has strong absorption at 405nm.
The supplied kit components are sufficient for performing up to 1000 assays in 96-well plate format, and are easily adaptable to cuvettes or 384-well plates.