ContactUsLinkComponent
Nalgene® Mini Hose Barb Connectors; PP, Thermo Scientific
Nalgene® Mini Hose Barb Connectors; PP, Thermo Scientific
Catalog # 62860-554
undefined
Nalgene® Mini Hose Barb Connectors; PP, Thermo Scientific
Catalog # 62860-554
Supplier Number:  7210-0500
Specifications
  • Environmentally Preferable:
  • Cat. No.:
    62860-554
  • Supplier no.:
    7210-0500
Specifications
About this item
Nalgene Mini Hose Barb Polypropylene Connectors allow connections between 3/4" sanitary fitting to flexible tubing
CAUTION! Plastic tanks are generally subject to more severe conditions than plastic labware; exposure is constant, stresses are greater and different classes and concentrations of chemicals are involved. Please pay special attention to chemical compatibility.
Comes with an inner diameter of 1/2".