Order Entry
United States
ContactUsLinkComponent
Geno-DNA-Template™ Yeast DNA Extraction Kit, G-Biosciences
Geno-DNA-Template™ Yeast DNA Extraction Kit, G-Biosciences
Catalog # 82021-534
Supplier:  G-Biosciences
CAS Number:  
undefined
Geno-DNA-Template™ Yeast DNA Extraction Kit, G-Biosciences
Catalog # 82021-534
Supplier:  G-Biosciences
Supplier Number:  786-134
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Application:
  • Method / Format:
    Precipitation
  • Prep size:
    Scalable
  • Refine target molecule:
    Total DNA
  • Sample type:
    Yeast/Fungi
  • Target molecule:
    DNA
  • Description:
    Yeast Geno-DNA-Template™
  • Size:
    100 Preps
  • Environmentally Preferable:
  • Description:
    Yeast Geno-DNA-Template™
  • Cat. No.:
    82021-534
  • Supplier no.:
    786-134

Specifications

About this item

G-Biosciences' Yeast Geno-DNA-Template™ extraction kit isolates high quality genomic DNA from yeast cultures

G-Biosciences' kit is based on a two-step lysis of yeast cells, using LongLife™ Zymolyase® and LongLife™ Proteinase K, followed by the removal of proteins and other cellular impurities by precipitation and centrifugation. The clean, genomic DNA is collected by precipitation and the high yield of extracted DNA has an average 100 kb length and has A₂₆₀/A₂₈₀ 1.8-2.0.

G-Biosciences' Yeast Geno-DNA-Template™ kit is supplied with reagents for extracting DNA from 100 x 1.5 mL yeast cultures.

The kit is compatible with PCR and other downstream applications.

Ordering information: Accessories available.