Order Entry
United States
Orders LinkContactUsLinkComponent
 

2-(2-Ethoxyethoxy)ethyl acetate 99%

2-(2-Ethoxyethoxy)ethyl acetate 99%
Certificates
2-(2-Ethoxyethoxy)ethyl acetate 99%

3 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Purity: 99% CAS No.: 112-15-2 Molecular Formula: CH3CO2(CH2CH2O)2C2H5 Formula Weight: 176.21 Flash Point: 95°(203°F) Refractive Index: 1.4210 Melting Point: -25° Boiling Point: 218-219° Beilstein Reference: 1764643 Merck Reference: 14,1800 MDL No.: MFCD00041928

Synonyms:  Diethylene glycol monoethyl ether acetate
  • CAS Number:112-15-2
  • Formula:C₈H₁₆O₄
  • MDL Number:MFCD00041928
  • Molecular Weight:176.21 g/mol
  • Merck Index:14,1800
  • Boiling Pt:218…219 °C
  • Melting Pt:–25 °C
  • Density:1.012
  • Flash Pt:95 °C(203 °F)
  • Storage Temperature:Ambient
  • EINECS:203-940-1
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • bottle
    100 ml
    Log in for availability
    Log in for availability
    Each (100ml)
    Each (100ml) - $20.50
    $20.50
    Each (100ml)
       $20.50

    Supplier Number: L13446-AE
  • bottle
    500 ml
    Log in for availability
    Log in for availability
    Each (500ml)
    Each (500ml) - $61.33
    $61.33
    Each (500ml)
       $61.33

    Supplier Number: L13446-AP
  • bottle
    2.5 L
    Log in for availability
    Log in for availability
    Each (2,500ml)
    Each (2,500ml) - $147.00
    $147.00
    Each (2,500ml)
       $147.00

    Supplier Number: L13446-0F

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.