


Hydrogen peroxide 27% (w/w) in aqueous solution stabilized
Catalog # AAAL13235-AP
Supplier: Thermo Scientific Chemicals
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:bottle
- Size:500 mL
- Cat. No.:AAAL13235-AP
- Supplier no.:L13235-AP
Specifications
About this item
:
Merck Index:14,4798
Boiling Pt:107 °C
Melting Pt:–26 °C
Density:1.100
Storage Temperature:Ambient
EINECS:231-765-0
UN:2014
ADR:"5.1",II
:34.01
:GHS03
:GHS05
:GHS07
CAS Number:7722-84-1
Formula:H₂O₂
MDL Number:MFCD00011333
ADR:5.1,II