Order Entry
United States
Orders LinkContactUsLinkComponent
1,2,6-Hexanetriol 96%

1,2,6-Hexanetriol 96%

1,2,6-Hexanetriol 96%
Certificates
1,2,6-Hexanetriol 96%
1,2,6-Hexanetriol 96%

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Purity: 96% CAS No.: 106-69-4 Molecular Formula: C6H14O3 Formula Weight: 134.18 Flash Point: 191°(375°F) Refractive Index: 1.4760 Melting Point: -20° Boiling Point: 166-168°/2mm Beilstein Reference: 1304479 MDL No.: MFCD00002976

Synonyms:  1,2,6-Trihydroxyhexane
  • CAS Number:106-69-4
  • Formula:C₆H₁₄O₃
  • MDL Number:MFCD00002976
  • Molecular Weight:134.18 g/mol
  • Boiling Pt:166…168 °C (2 mmHg)
  • Boiling Pt:166…168 °C/2 mm
  • Melting Pt:–20 °C
  • Density:1.106
  • Flash Pt:191 °C (375 °F)
  • Flash Pt:191 °C(375 °F)
  • Storage Temperature:Ambient
  • EINECS:203-424-6

Documents

Product Family Options

Product Information

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.