Order Entry
United States
ContactUsLinkComponent
3-Mercaptobutanone 98% stabilized
Flammable
Irritant
3-Mercaptobutanone 98% stabilized
Catalog # AAB22494-14
CAS Number:  
Flammable
Irritant
3-Mercaptobutanone 98% stabilized
Catalog # AAB22494-14
Supplier Number:  B22494-14
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    bottle
  • Size:
    25 g
  • Cat. No.:
    AAB22494-14
  • Supplier no.:
    B22494-14

Specifications

About this item

 :
 :104.17 g/mol
Boiling Pt:50 °C/18 mm
Density:1.015
Flash Pt:47 °C (117 °F)
Storage Temperature:Refrigerator
EINECS:255-082-2
UN:1224
ADR:3,III
 :Stabilised with calcium carbonate 0.1%
 :GHS02
 :GHS07
CAS Number:40789-98-8
Formula:C₄H₈OS
MDL Number:MFCD00085206