Bis(2-methoxyethyl)ether 99% stabilised
About this item
Purity: 99% CAS No.: 111-96-6 Molecular Formula: C6H14O3 Formula Weight: 134.18 Flash Point: 51°(124°F) Refractive Index: 1.4080 Odor: Ether-like Melting Point: -64° Boiling Point: 161-162° Beilstein Reference: 1736101 Merck Reference: 14,3165 UN No.: 3271 MDL No.: MFCD00008503
- CAS Number:111-96-6
- Formula:C₆H₁₄O₃
- MDL Number:MFCD00008503
- Molecular Weight:134.18 g/mol
- Merck Index:14,3165
- Boiling Pt:161…162 °C
- Melting Pt:–64 °C
- Density:0.944
- Flash Pt:51 °C (124 °F)
- Storage Temperature:Ambient
- UN:3271
- ADR:3,III
- :Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 100 ppm
- EINECS:203-924-4
1 Options Available Below
Product Details & Documents
Purity: 99% CAS No.: 111-96-6 Molecular Formula: C6H14O3 Formula Weight: 134.18 Flash Point: 51°(124°F) Refractive Index: 1.4080 Odor: Ether-like Melting Point: -64° Boiling Point: 161-162° Beilstein Reference: 1736101 Merck Reference: 14,3165 UN No.: 3271 MDL No.: MFCD00008503
- CAS Number:111-96-6
- Formula:C₆H₁₄O₃
- MDL Number:MFCD00008503
- Molecular Weight:134.18 g/mol
- Merck Index:14,3165
- Boiling Pt:161…162 °C
- Melting Pt:–64 °C
- Density:0.944
- Flash Pt:51 °C (124 °F)
- Storage Temperature:Ambient
- UN:3271
- ADR:3,III
- :Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 100 ppm
- EINECS:203-924-4
Product Family Options
Product Information
- Pack typeSizeAvailabilityPrice
- Supplier Number: A13397-AP
- bottle2.5 LDiscontinued-Supplier Number: -
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



