
Nitric acid standardised solution 2.0 N
Supplier: Thermo Scientific Chemicals
Certificates
About this item
CAS Number:7697-37-2
:
Formula:HNO₃
MDL Number:MFCD00011349
:63.01
Density:1.06
Storage Temperature:Ambient
EINECS:231-714-2
UN:3264
ADR:8,II
:GHS05
Related Information
Certificates
Search by Lot NumberAlfa Aesar_Buy 3 Get 2 Complimentary items
Promotion Material
VWR GHS Compliance Letter (US)
Additional Product Details
Frequently Bought Together