ContactUsLinkComponent
Nalgene® Sterile Specimen Cryogenic Vial with Screw Closure, Thermo Scientific
Nalgene® Sterile Specimen Cryogenic Vial with Screw Closure, Thermo Scientific
  66008-740
undefined
Nalgene® Sterile Specimen Cryogenic Vial with Screw Closure, Thermo Scientific
  66008-740
 :  5005-0015
  • Cap included:
    Yes
  • Capacity:
    15.00 ml
  • Cat. No.:
    66008-740
  • Diameter ×H:
    33×47 mm
  • Supplier no.:
    5005-0015
Nalgene® Sterile Specimen Cryogenic Vials are designed for storage of solid specimens.
They feature a linerless closure, as well as a wide-mouth, shoulderless vial that allows easy access to sample with forceps. The vials are for use in mechanical freezers only.
 : This product is in compliance to Directive 98/79/EC of the European Parliament and of the Council of 27 October 1998 on in vitro diagnostic medical devices. Certified non-cytotoxic and non-pyrogenic.
 : Packaged 5 vials per easy open bag.