Nalgene® Economy Polypropylene Graduated Cylinders, Thermo Scientific
Nalgene® Economy PP Graduated Cylinders exhibit excellent chemical resistance and have a generous pour spout for easily dispensing liquids
They have large rounded bases and are ribbed for reinforcement and stability.The cylinders are calibrated TC and TD at 20°C. Also they feature good temperature resistance up to 135°C.
- Catalog No:
- 24780-265
- 24780-287
- 24780-268
- 24780-323
- 24780-301
- 24780-367
- 24780-345
- Form:
- Tall
- Tall
- Tall
- Tall
- Tall
- Tall
- Tall
- Environmentally Preferable:
- Low Manufacturing Impact, Product Transparency and Disclosure
- Low Manufacturing Impact, Product Transparency and Disclosure
- Low Manufacturing Impact, Product Transparency and Disclosure
- Low Manufacturing Impact, Product Transparency and Disclosure
- Low Manufacturing Impact, Product Transparency and Disclosure
- Product Transparency and Disclosure
- Low Manufacturing Impact, Product Transparency and Disclosure
- Environmental Certification:
- ISO14001 (Certificate Registration No. EMS 752603, ACT Label
- ISO14001 (Certificate Registration No. EMS 752603), ACT label
- ISO14001 (Certificate Registration No. EMS 752603, ACT Label
- ISO14001 (Certificate Registration No. EMS 752603), ACT label
- ISO14001 (Certificate Registration No. EMS 752603), ACT label
- ACT Label
- ISO14001 (Certificate Registration No. EMS 752603), ACT label
Frequently Bought Together