VITLAB® Reagent Bottles, PP with NS Stoppers, BrandTech
Catalog # 83008-524
Supplier: Brandtech
Specifications
- [ST] joint no.:29/32
- Capacity:1 L (33.8 oz.)
- Cat. No.:83008-524
- Diameter ×H:108×213 mm
- Supplier no.:V100694
Specifications
About this item
Slope-shouldered polypropylene bottles feature NS tapered polypropylene stoppers to help prevent leakage.
- Break-resistant
- Polypropylene bottle and stopper for high chemical resistance
- High translucency
- Rugged design
5 liter size includes a handle to facilitate movement.