ContactUsLinkComponent
Rodent Restrainer, Plas-Labs™
Rodent Restrainer, Plas-Labs™
  95042-916
 :  Plas-Labs
cUL Listed
CSA Certified
Small Business Enterprise
UL Listed
Rodent Restrainer, Plas-Labs™
  95042-916
 :  Plas-Labs
 :  530-POR
cUL Listed
CSA Certified
Small Business Enterprise
UL Listed
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Description:
    Guinea pigs restrainer for <750 g
  • Cat. No.:
    95042-916
  • Supplier no.:
    530-POR
The PLAS LABS 530-POR, Bowman Style Rodent Restrainer has been re-engineered and refined to accommodate larger sized animals up to 750 grams
The positions of the stainless steel rods around the rodent provide easy access for many types of research applications, especially obesity studies.