SureBlue™ TMB Microwell Substrate develops a deep blue color in the presence of peroxidase-labeled conjugates measurable at 650 nm and turns yellow when stopped by acidification and read at 450 nm
Recommended for ELISA. Single component, ready to use format of 3,3',5,5' - Tetramethylbenzidine. No reagent preparation required, very convenient.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Host:
- Primary antibody reactivity:
- Environmentally Preferable: