Nalgene® Narrow-Mouth Packaging Bottles, PPCO with Closures, Bulk Pack, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
These narrow mouth polypropylene copolymer bottles have a robust construction for packaging use and are featured with polypropylene closures.
- Exhibit excellent impact and chemical resistance and clarity
- Leak proof and autoclavable and have a translucent color
Certifications: Manufactured from validated and controlled resins meeting a variety of food and pharmaceutical regulatory requirements.
Ordering information: They are bulk packed for packaging applications.
Specifications
- Catalog No:
- 16059-524
- 16059-534
- 16059-526
- 16059-528
- 16059-530
- 16059-532
- Environmentally Preferable:
- Low Manufacturing Impact
- Environmental Certification:
- ISO14001 (Certificate Registration No. EMS 752603)