Order Entry
United States
Orders LinkContactUsLinkComponent
D-(-)-N-Methylglucamine

D-(-)-N-Methylglucamine

Bottle, 500 mg
Catalog # 101095-412
Marketsource
Supplier:   USP
Supplier #:  1379140
D-(-)-N-Methylglucamine
Catalog # 101095-412
Supplier:  USP
Marketsource
CAS Number:  6284-40-8
D-(-)-N-Methylglucamine
D-(-)-N-Methylglucamine

Bottle, 500 mg

CAS Number:  6284-40-8
Price
$407.69
Log in for availability
Each (500mg)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  Meglumine, (-)-1-Deoxy-1-(methylamino)-D-glucitol, N-Methyl-D-glucamine, (-)-N-Methyl-D-glucamine, (2R,3R,4R,5S)-6-(Methylamino)hexane-1,2,3,4,5-pentaol, (-)-1-Methylamino-1-deoxy-D-glucitol
  • CAS Number:6284-40-8
  • Formula:H₃CNHCH₂(CH(OH))₄CH₂OH
  • MDL Number:MFCD00004707
  • Molecular Weight:195.22 g/mol
  • Merck Index:12,06154
  • Melting Pt:128…131 °C
  • Storage Temperature:Ambient
  • EINECS:228-506-9

Documents

Specifications

Specifications

  • Pack type:
    Bottle
  • Size:
    500 mg

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.