Order Entry
United States
Orders LinkContactUsLinkComponent
Lanolin stabilised

Lanolin stabilised

Bottle, 20 g
Catalog # 101095-306
Marketsource
Supplier:   USP
Supplier #:  1356880
Lanolin stabilised
Catalog # 101095-306
Supplier:  USP
Marketsource
CAS Number:  8006-54-0
Lanolin stabilised
Lanolin stabilised

Bottle, 20 g

CAS Number:  8006-54-0
Price
$438.46
Log in for availability
Each (20g/mol)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  Wool fat
  • CAS Number:8006-54-0
  • MDL Number:MFCD00081740
  • Merck Index:12,05371
  • Melting Pt:38…44 °C
  • Density:0.932…0.945 g/cm³ (15 °C)
  • Flash Pt:113 °C (closed cup)
  • EINECS:232-348-6

Documents

Specifications

Specifications

  • Pack type:
    Bottle
  • Size:
    20 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.