Order Entry
United States
Orders LinkContactUsLinkComponent
Capsaicin

Capsaicin

Bottle, 100 mg
Catalog # 101094-486
Marketsource
Supplier:   USP
Supplier #:  1091108
Capsaicin
Catalog # 101094-486
Supplier:  USP
Marketsource
CAS Number:  404-86-4
Capsaicin
Capsaicin

Bottle, 100 mg

CAS Number:  404-86-4
Price
$458.46
Log in for availability
Each (100mg)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  8-Methyl-N-vanillyl-6-none-amid
  • CAS Number:404-86-4
  • Formula:C₁₈H₂₇NO₃
  • MDL Number:MFCD00017259
  • Molecular Weight:305.42 g/mol
  • Merck Index:12,01811
  • Boiling Pt:210…220 °C (0.01 mmHg)
  • Melting Pt:66.1 °C
  • EINECS:206-969-8
  • UN:2811
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Bottle
  • Size:
    100 mg

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.