Potassium bromide, crystal rectangle for spectrophotometry, unpolished, drilled
Catalog # 100212-952
Supplier: International Crystal Labs
Minority or Woman-Owned Business Enterprise
CAS Number:
Specifications
- Size:38.5×19.5×4
- Environmentally Preferable:
- Cat. No.:100212-952
- Supplier no.:0002N-178
Specifications
About this item
:
Merck Index:13,07701
Boiling Pt:1435 °C (1013 hPa)
Melting Pt:730 °C
Density:2.75 g/cm³ (20 °C)
:119 g/mol
Storage Temperature:Ambient
EINECS:231-830-3
:GHS07
CAS Number:7758-02-3
Formula:KBr
MDL Number:MFCD00011358