Potassium bromide, crystal disk for spectrophotometry, unpolished, drilled
Catalog # 100211-346
Supplier: International Crystal Labs
Minority or Woman-Owned Business Enterprise
CAS Number:
Specifications
- Size:32×3 mm
- Environmentally Preferable:
- Cat. No.:100211-346
- Supplier no.:0002N-534
Specifications
About this item
:
Merck Index:13,07701
Boiling Pt:1435 °C (1013 hPa)
Melting Pt:730 °C
Density:2.75 g/cm³ (20 °C)
:119 g/mol
Storage Temperature:Ambient
EINECS:231-830-3
:GHS07
CAS Number:7758-02-3
Formula:KBr
MDL Number:MFCD00011358