Zinc sulfide, crystal disk for spectrophotometry, polished
Supplier: International Crystal Labs
Certificates
About this item
CAS Number:1314-98-3
:
Formula:SZn
MDL Number:MFCD00011301
:97.46 g/mol
Merck Index:12,10294
Melting Pt:1064 °C
Density:4.1 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-251-3
Frequently Bought Together