Order Entry
United States
ContactUsLinkComponent
Potassium bromide, crystal disk for spectrophotometry, polished, drilled
Irritant
Potassium bromide, crystal disk for spectrophotometry, polished, drilled
Catalog # 100212-568
Minority or Woman-Owned Business Enterprise
CAS Number:  
undefined
Irritant
Potassium bromide, crystal disk for spectrophotometry, polished, drilled
Catalog # 100212-568
Supplier Number:  0002C-118
Minority or Woman-Owned Business Enterprise
CAS Number:  

Specifications

  • Size:
    32×3 mm
  • Cat. No.:
    100212-568
  • Supplier no.:
    0002C-118

Specifications

About this item

 :
Merck Index:13,07701
Boiling Pt:1435 °C (1013 hPa)
Melting Pt:730 °C
Density:2.75 g/cm³ (20 °C)
 :119 g/mol
Storage Temperature:Ambient
EINECS:231-830-3
 :GHS07
CAS Number:7758-02-3
Formula:KBr
MDL Number:MFCD00011358