Potassium chloride, crystal rectangle for spectrophotometry, polished, drilled
Supplier: International Crystal Labs
Certificates
About this item
CAS Number:7447-40-7
:
Formula:KCl
MDL Number:MFCD00011360
:74.55 g/mol
Merck Index:14,07621
Boiling Pt:1420 °C (1013 hPa)
Melting Pt:773 °C
Density:1.98 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-211-8
Frequently Bought Together