



Trimethylamine 24% in aqueous solution
Supplier: Spectrum Chemical Mfg. Corp.
Certificates
About this item
CAS Number:75-50-3
:
Formula:C₃H₉N
MDL Number:MFCD00008327
Merck Index:12,09841
Boiling Pt:2.87 °C (1013 hPa)
Melting Pt:–117.1 °C
Density:0.932
Flash Pt:–20 °C
UN:1297
ADR:38,II
:59.11 g/mol
EINECS:200-875-0
:GHS02
:GHS04
:GHS05
:GHS07
Storage Temperature:Refrigerator
Related Information
Certificates
Search by Lot NumberT2272_spec.pdf
Product specification
VWR GHS Compliance Letter (US)
Additional Product Details
Frequently Bought Together