Order Entry
United States
Orders LinkContactUsLinkComponent
1,2,6-Hexanetriol ≥97% for synthesis

1,2,6-Hexanetriol ≥97% for synthesis

1,2,6-Hexanetriol ≥97% for synthesis
Certificates

4 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

1,2,6-Trihydroxyhexane is a stable, high boiling, liquid polyol that is miscible with water and is non-toxic. 1,2,6-TRIHYDROXYHEXANE is used in cosmetic and pharmaceutical cream formulations as a moisturizing agent (humectant). Its excellent solvent properties also lend it useful in as a carrier solvent for ink and paint formulations, as well as in photographic and textile formulations. Due to the interesting physical and chemical cross-linking properties of this triol, it finds utility in the manufacture of synthetic resins, alkyd resins, plasticizers and polyurethane systems.

Synonyms:  1,2,6-Trihydroxyhexane
  • CAS Number:106-69-4
  • Formula:C₆H₁₄O₃
  • MDL Number:MFCD00002976
  • Molecular Weight:134.18 g/mol
  • Boiling Pt:323 °C (1013 hPa)
  • Melting Pt:25…32 °C
  • Density:1.109
  • Storage Temperature:Ambient
  • EINECS:203-424-6

Documents

Specifications for Product Family

Specification Test Results

  • Assay:
    97% Min.
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Glass, amber
    25 g
    Log in for availability
    Log in for availability
    Each (25g)
    Each (25g) - $112.95
    $112.95
    Each (25g)
       $112.95

    Supplier Number: T2117-25GM
  • Glass, amber
    100 g
    Log in for availability
    Log in for availability
    Each (100g)
    Each (100g) - $289.06
    $289.06
    Each (100g)
       $289.06

    Supplier Number: T2117-100GM
  • Glass, amber
    500 g
    Log in for availability
    Log in for availability
    Each (500g)
    Each (500g) - $1,121.75
    $1,121.75
    Each (500g)
       $1,121.75

    Supplier Number: T2117-500GM
  • Glass, amber
    1 kg
    Log in for availability
    Log in for availability
    Each (1kg)
    Each (1kg) - $1,941.45
    $1,941.45
    Each (1kg)
       $1,941.45

    Supplier Number: T2117-1KG

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.