Order Entry
United States
Orders LinkContactUsLinkComponent
Sorbitan monooleate NF suitable for use as excipient

Sorbitan monooleate NF suitable for use as excipient

Sorbitan monooleate NF suitable for use as excipient
Certificates
Sorbitan monooleate NF suitable for use as excipient

6 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Synonyms:  Sorbitan oleate, Span 80, (2R,3R,4R,5S)-Hexane-1,2,3,4,5,6-hexaol mono-(9Z)-9-octadecenoate
  • CAS Number:1338-43-8
  • Formula:C₂₄H₄₄O₆
  • MDL Number:MFCD00080948
  • Molecular Weight:428.61 g/mol
  • Density:0.986
  • Storage Temperature:Ambient
  • EINECS:215-665-4

Documents

Certificates
Search by Lot Number
Specifications for Product Family

Specification Test Results

  • Assay:
    72% Min.

Specifications

Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Glass, amber
    500 ml
    Log in for availability
    Log in for availability
    Each (500ml)
    Each (500ml) - $183.81
    $183.81
    Each (500ml)
       $183.81

    Supplier Number: SPA83-500MLG
    Specifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    700011-080
    Small Business Enterprise
  • Glass, amber
    500 ml
    Log in for availability
    Log in for availability
    Each (500ml)
    Each (500ml) - $224.97
    $224.97
    Each (500ml)
       $224.97

    Supplier Number: SPA83-500MLG
    Specifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    95036-010
    Small Business Enterprise
  • Glass, amber
    4 L
    Log in for availability
    Log in for availability
    Each (4l)
    Each (4l) - $615.83
    $615.83
    Each (4l)
       $615.83

    Supplier Number: SPA83-4LTGL
    Specifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    95036-008
    Small Business Enterprise
  • Poly drum
    20 L
    Log in for availability
    Log in for availability
    Each (20l)
    Each (20l) - $1,748.40
    $1,748.40
    Each (20l)
       $1,748.40

    Supplier Number: SPA83-20LT
    Specifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    89079-030
    Small Business Enterprise
  • Drum
    200 L
    Log in for availability
    Log in for availability
    Each (200l)
    Each (200l) - $5,363.39
    $5,363.39
    Each (200l)
       $5,363.39

    Supplier Number: SPA83-200LTB
    Specifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    700011-078
    Small Business Enterprise
  • Drum
    200 L
    Log in for availability
    Log in for availability
    Each (200l)
    Each (200l) - $5,273.74
    $5,273.74
    Each (200l)
       $5,273.74

    Supplier Number: SPA83-200LTB
    Specifications:
    More
    Specifications
    Cat. No.
    89079-032

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.