Sorbitan tristearate for synthesis
About this item
Sorbitan Tristearate is a triester of stearic acid and hexitol anhydrides derived from sorbitol. It functions as a surfactant and emulsifying agent. This product appears as a white to tan-colored solid. In cosmetics and personal care products, Sorbitan Esters are used in a variety of products including skin care products, skin cleansing products, moisturizers, eye makeup and other makeup
- CAS Number:26658-19-5
- Formula:C₆₀H₁₁₄O₈
- MDL Number:MFCD00151165
- Molecular Weight:963.56 g/mol
- Storage Temperature:Ambient
- EINECS:247-891-4
2 Options Available Below
Product Details & Documents
Sorbitan Tristearate is a triester of stearic acid and hexitol anhydrides derived from sorbitol. It functions as a surfactant and emulsifying agent. This product appears as a white to tan-colored solid. In cosmetics and personal care products, Sorbitan Esters are used in a variety of products including skin care products, skin cleansing products, moisturizers, eye makeup and other makeup
- CAS Number:26658-19-5
- Formula:C₆₀H₁₁₄O₈
- MDL Number:MFCD00151165
- Molecular Weight:963.56 g/mol
- Storage Temperature:Ambient
- EINECS:247-891-4
Product Family Options
Product Information
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



