Order Entry
United States
Orders LinkContactUsLinkComponent
Sodium hydroxide, coarse granules, Technical Grade suitable for use as excipient

Sodium hydroxide, coarse granules, Technical Grade suitable for use as excipient

Sodium hydroxide, coarse granules, Technical Grade suitable for use as excipient
Certificates
Sodium hydroxide, coarse granules, Technical Grade suitable for use as excipient

3 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Sodium Hydroxide, Coarse Granular, Technical is a strong base that is also used in manufacturing of sodium salts and detergents, pH regulation, and organic synthesis. Technical grade products supplied by Spectrum are indicative of a grade suitable for general industrial use. These materials may or may not have a Certificate of Analysis available

Synonyms:  Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye
  • CAS Number:1310-73-2
  • Formula:NaOH
  • MDL Number:MFCD00003548
  • Molecular Weight:40 g/mol
  • Merck Index:14,08627
  • Boiling Pt:1390 °C (1013 hPa)
  • Melting Pt:323 °C
  • Density:2.13 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-185-5
  • UN:1823
  • ADR:8,II
  • REACH:01-2119457892-27

Documents

Certificates
Search by Lot Number
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Poly bottle
    500 g
    Log in for availability
    Log in for availability
    Each (500g)
    Each (500g) - $111.36
    $111.36
    Each (500g)
       $111.36
    Minimum order quantity is 1

    Supplier Number: S1302-500GM
  • Poly bottle
    2.5 kg
    Log in for availability
    Log in for availability
    Each (2,500g)
    Each (2,500g) - $190.42
    $190.42
    Each (2,500g)
       $190.42
    Minimum order quantity is 1

    Supplier Number: S1302-2.5KG
  • Poly pail
    12 kg
    Log in for availability
    Log in for availability
    Each (12kg)
    Each (12kg) - $598.03
    $598.03
    Each (12kg)
       $598.03

    Supplier Number: S1302-12KG

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.