Potassium hydroxide 45% (w/w) in aqueous solution, Reagent Grade
About this item
Potassium Hydroxide, 45 Percent (w/w) Aqueous Solution, Reagent is an aqueous solution of Caustic Potash and is the protypical strong base. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.
- CAS Number:1310-58-3
- Formula:KOH
- MDL Number:MFCD00003553
- Molecular Weight:56.11 g/mol
- Storage Temperature:Ambient
- EINECS:215-181-3
- UN:1814
- ADR:8,II
4 Options Available Below
Product Details & Documents
Potassium Hydroxide, 45 Percent (w/w) Aqueous Solution, Reagent is an aqueous solution of Caustic Potash and is the protypical strong base. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.
- CAS Number:1310-58-3
- Formula:KOH
- MDL Number:MFCD00003553
- Molecular Weight:56.11 g/mol
- Storage Temperature:Ambient
- EINECS:215-181-3
- UN:1814
- ADR:8,II
Documents
Certificates
Search by Lot NumberP1320_spec.pdf
Product specification
VWR GHS Compliance Letter (US)
Additional Product Details
Product Family Options
Product Information
- Pack typeSizeAvailabilityPrice
- Supplier Number: P1320-500MLP
- Supplier Number: P1320-4LTPL
- Supplier Number: P1320-20LT
- Supplier Number: P1320-200LTB
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



