Order Entry
United States
Orders LinkContactUsLinkComponent
Methyl violet 2B for synthesis

Methyl violet 2B for synthesis

Corrosive
Health
Irritant
Environment
Methyl violet 2B for synthesis
Certificates
Methyl violet 2B for synthesis

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Methyl Violet 2B, also known as crystal violet, can be used as a purple dye for textiles and to give deep violet colors in paint and ink.

Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Synonyms:  Basic Violet 1, 4-[4,4'-Bis(dimethylamino) benzhydrylidene]cyclohexa-2,5-dien-1-ylidene]methylammonium chloride
  • CAS Number:8004-87-3
  • Formula:C₂₄H₂₈ClN₃
  • MDL Number:MFCD00012568
  • Molecular Weight:393.96 g/mol
  • Melting Pt:137 °C
  • Storage Temperature:Ambient
  • UN:3077
  • ADR:9,III

Documents

Certificates
Search by Lot Number
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Glass, amber
    25 g
    Log in for availability
    Log in for availability
    Each (25g)
    Each (25g) - $111.37
    $111.37
    Each (25g)
       $111.37

    Supplier Number: ME175-25GM
  • Glass, amber
    100 g
    Log in for availability
    Log in for availability
    Each (100g)
    Each (100g) - $352.35
    $352.35
    Each (100g)
       $352.35

    Supplier Number: ME175-100GM

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.