Lanolin USP suitable for use as excipient
About this item
All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
- CAS Number:8006-54-0
- MDL Number:MFCD00081740
- Merck Index:12,05371
- Melting Pt:38…44 °C
- Density:0.932…0.945 g/cm³ (15 °C)
- Flash Pt:113 °C (closed cup)
- Storage Temperature:Ambient
- EINECS:232-348-6
5 Options Available Below
Product Details & Documents
All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
- CAS Number:8006-54-0
- MDL Number:MFCD00081740
- Merck Index:12,05371
- Melting Pt:38…44 °C
- Density:0.932…0.945 g/cm³ (15 °C)
- Flash Pt:113 °C (closed cup)
- Storage Temperature:Ambient
- EINECS:232-348-6
Documents
Product Family Options
Product Information
- Pack typeSizeAvailabilityPrice
- Supplier Number: LA109-430GM
- Supplier Number: LA109-430CS6
- Supplier Number: LA109-18KGBL
- Supplier Number: LA109-180KGB
- Supplier Number: LA109-180KGB
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



