Order Entry
United States
ContactUsLinkComponent
Lithium hydroxide, anhydrous
Toxic
Corrosive
Lithium hydroxide, anhydrous
Catalog # 700000-916
Small Business Enterprise
CAS Number:  
Toxic
Corrosive
Lithium hydroxide, anhydrous
Catalog # 700000-916
Supplier Number:  L1121-50KGBL
Small Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Fiber drum
  • Size:
    50 kg
  • Cat. No.:
    700000-916
  • Supplier no.:
    L1121-50KGBL

Specifications

About this item

 :
 :23.95 g/mol
Merck Index:13,05556
Boiling Pt:924 °C (1013 hPa)
Melting Pt:462 °C
Density:1.46 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-183-4
UN:2680
ADR:8,II
 :GHS06
 :GHS05
CAS Number:1310-65-2
Formula:LiOH
MDL Number:MFCD00011095