Collodion USP
About this item
Collodion, USP is used as a surgical dressing. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities
- CAS Number:9004-70-0
- MDL Number:MFCD00081525
- Merck Index:13,02508
- Boiling Pt:∼34 °C (1013 hPa)
- Melting Pt:160 °C
- Density:0.75 g/cm³ (20 °C)
- Flash Pt:–19 °C
- Storage Temperature:Ambient
- UN:2059
- ADR:3,II
4 Options Available Below
Product Details & Documents
Collodion, USP is used as a surgical dressing. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities
- CAS Number:9004-70-0
- MDL Number:MFCD00081525
- Merck Index:13,02508
- Boiling Pt:∼34 °C (1013 hPa)
- Melting Pt:160 °C
- Density:0.75 g/cm³ (20 °C)
- Flash Pt:–19 °C
- Storage Temperature:Ambient
- UN:2059
- ADR:3,II
Specifications for Product Family
Specification Test Results
- Assay:5% Min.
Product Family Options
Product Information
- Pack typeSizeAvailabilityPrice
- Supplier Number: CO120-100MLG
- Glass, amber100 mlDiscontinued-Supplier Number: -
- Supplier Number: CO120-500MLG
- Glass, amber500 mlDiscontinued-Supplier Number: -
- Supplier Number: CO120-4LTGL
- Supplier Number: CO120-200LTB
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



