Butyl acetate ≥99.5% ACS for synthesis
Supplier: Spectrum Chemical Mfg. Corp.
About this item
Synonyms: Butyl ethanoate, Acetic acid butyl ester, Acetic acid n-butyl ester, n-Butyl acetate
- CAS Number:123-86-4
- Formula:CH₃COO(CH₂)₃CH₃
- MDL Number:MFCD00009445
- Molecular Weight:116.16 g/mol
- Merck Index:13,01534
- Boiling Pt:126 °C (1013 hPa)
- Melting Pt:–76 °C
- Density:0.882
- Flash Pt:25 °C
- Storage Temperature:Ambient
- EINECS:204-658-1
- UN:1123
- ADR:3,II
7 Options Available Below
Product Details & Documents
Synonyms: Butyl ethanoate, Acetic acid butyl ester, Acetic acid n-butyl ester, n-Butyl acetate
- CAS Number:123-86-4
- Formula:CH₃COO(CH₂)₃CH₃
- MDL Number:MFCD00009445
- Molecular Weight:116.16 g/mol
- Merck Index:13,01534
- Boiling Pt:126 °C (1013 hPa)
- Melting Pt:–76 °C
- Density:0.882
- Flash Pt:25 °C
- Storage Temperature:Ambient
- EINECS:204-658-1
- UN:1123
- ADR:3,II
Specifications for Product Family
Specification Test Results
- Assay:99.5% Min.
Specifications
Product Family Options
Product Information
- Pack typeSizeAvailabilityPrice
- Supplier Number: B1182-100MLSpecifications:
More
Specifications
Cat. No.700004-388 - Supplier Number: B1182-500MLGSpecifications:
More
Specifications
Cat. No.700004-400 - Supplier Number: B1182-1LTGLSpecifications:Small Business Enterprise
More
Specifications
Cat. No.700004-390Small Business Enterprise - Supplier Number: B1182-2.5LTPSpecifications:Small Business Enterprise
More
Specifications
Cat. No.700004-392Small Business Enterprise - Supplier Number: B1182-4LTGLSpecifications:
More
Specifications
Cat. No.700004-398 - Supplier Number: B1182-20LTMTSpecifications:
More
Specifications
Cat. No.700004-396 - Supplier Number: B1182-200LTBSpecifications:Small Business Enterprise
More
Specifications
Cat. No.700004-394Small Business Enterprise
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



