Order Entry
United States
Orders LinkContactUsLinkComponent
 

Arachidic acid ≥99% for synthesis

Arachidic acid ≥99% for synthesis
Certificates

4 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Arachidic Acid, also known as eicosanoic acid or n-eicosanoic acid, is a saturated fatty acid used in the production of lubricants and detergents.

Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Synonyms:  Eicosanylic acid, Arachidinic acid, Icosanoic acid, Eicosanoic acid
  • CAS Number:506-30-9
  • Formula:C₂₀H₄₀O₂
  • MDL Number:MFCD00002755
  • Molecular Weight:312.54 g/mol
  • Merck Index:12,00802
  • Boiling Pt:328 °C (1013 hPa)
  • Melting Pt:75.5 °C
  • Density:0.824 g/cm³ (100 °C) (lit.)
  • Storage Temperature:Ambient
  • EINECS:208-031-3
Specifications for Product Family

Specification Test Results

  • Assay:
    99% Min.
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Glass, amber
    1 g
    Log in for availability
    Log in for availability
    Each (1g)
    Each (1g) - $88.51
    $88.51
    Each (1g)
       $88.51

    Supplier Number: A3899-1GM
  • Glass, amber
    5 g
    Log in for availability
    Log in for availability
    Each (5g)
    Each (5g) - $254.62
    $254.62
    Each (5g)
       $254.62

    Supplier Number: A3899-5GM
  • Glass, amber
    10 g
    Log in for availability
    Log in for availability
    Each (10g)
    Each (10g) - $395.44
    $395.44
    Each (10g)
       $395.44

    Supplier Number: A3899-10GM
  • Glass, amber
    25 g
    Log in for availability
    Log in for availability
    Each (25g)
    Each (25g) - $757.89
    $757.89
    Each (25g)
       $757.89

    Supplier Number: A3899-25GM

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.