Tellurium dioxide ≥99.999% (Te basis), PURATREM
:7446-07-3
:
:O₂Te
:MFCD00011263
:159.6 g/mol
:14,09123
:450 °C (subl.)
:733 °C
:5.6 g/cm³ (20 °C)
:Ambient
:231-193-1
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.