Order Entry
United States
Orders LinkContactUsLinkComponent
Selenium dioxide ≥99.8%

Selenium dioxide ≥99.8%

 
 
 
Marketsource
Selenium dioxide ≥99.8%
Marketsource
 
Selenium dioxide ≥99.8%

 

 
   
 
  •  :7446-08-4
  •  :SeO₂
  •  :MFCD00003562
  •  :110.96 g/mol
  •  :13,08508
  •  :340…350 °C (subl.)
  •  :315 °C
  •  :3.95
  •  :Ambient
  •  :231-194-7
  •  :3283
  •  :6.1,III
Product Family Options

 

  • Pack type
    Size
     
     
  • Glass bottle
    100 g
       
       
    Each (100g)
    Each (100g) - $164.18
    $164.18
    Each (100g)
       $164.18

     : 93-3406-100G
     
    Marketsource
    Marketsource
  • Glass bottle
    500 g
       
       
    Each (500g)
    Each (500g) - $646.83
    $646.83
    Each (500g)
       $646.83

     : 93-3406-500G
     
    Marketsource
    Marketsource

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.