Phosphorus(V) chloride ≥98%
:10026-13-8
:
:Cl₅P
:MFCD00011439
:208.24 g/mol
:12,07508
:166.8 °C (subl.)
:3.6 g/cm³ (20 °C)
:233-060-3
:1806
:8,II
:GHS06
:GHS05
:GHS08
:Ambient
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.