Order Entry
United States
Orders LinkContactUsLinkComponent
Lead ≥99.9995%, wire

Lead ≥99.9995%, wire

Health
Irritant
Environment
Marketsource
Supplier:   Strem Chemicals Inc
Lead ≥99.9995%, wire
Marketsource
Certificates

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
  • CAS Number:7439-92-1
  • Formula:Pb
  • MDL Number:MFCD00134050
  • Molecular Weight:207.2 g/mol
  • Merck Index:12,05410
  • Boiling Pt:1620 °C
  • Melting Pt:327 °C
  • Density:11.288
  • Storage Temperature:Ambient
  • EINECS:231-100-4
Specifications for Product Family

Specifications

Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Glass bottle
    2 m
    Log in for availability
    Log in for availability
    Each (2m)
    Each (2m) - $239.20
    $239.20
    Each (2m)
       $239.20

    Supplier Number: 82-0200-2MT
    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    100205-214
    Marketsource
  • Glass bottle
    10 m
    Log in for availability
    Log in for availability
    Each (10m)
    Each (10m) - $959.63
    $959.63
    Each (10m)
       $959.63

    Supplier Number: 82-0200-10MT
    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    100208-974
    Marketsource

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.