Order Entry
United States
Orders LinkContactUsLinkComponent
 

Platinum ≥99.99%, wire

Marketsource
Supplier:   Strem Chemicals Inc
Platinum ≥99.99%, wire
Marketsource
Certificates
Platinum ≥99.99%, wire

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  Platinium
  • CAS Number:7440-06-4
  • Formula:Pt
  • MDL Number:MFCD00011179
  • Molecular Weight:195.08 g/mol
  • Merck Index:12,07689
  • Boiling Pt:3827 °C
  • Melting Pt:1769 °C
  • Density:21.45
  • Storage Temperature:Ambient
  • EINECS:231-116-1
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Glass bottle
    1 m
    Log in for availability
    Log in for availability
    Each (1m)
    Each (1m) - $208.06
    $208.06
    Each (1m)
       $208.06

    Supplier Number: 78-0080-1MT
    Specifications:
    Marketsource
    Marketsource
  • Glass bottle
    5 m
    Log in for availability
    Log in for availability
    Each (5m)
    Each (5m) - $818.09
    $818.09
    Each (5m)
       $818.09

    Supplier Number: 78-0080-5MT
    Specifications:
    Marketsource
    Marketsource

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.