ContactUsLinkComponent
Lysine-C™, Sequencing Grade, G-Biosciences
Lysine-C™, Sequencing Grade, G-Biosciences
Catalog # 75998-072
Supplier:  G-Biosciences
CAS Number:  
Lysine-C™, Sequencing Grade, G-Biosciences
Catalog # 75998-072
Supplier:  G-Biosciences
Supplier Number:  786-14
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    SG-Lysine-C™
  • Size:
    2 x 5 μg
  • Cat. No.:
    75998-072
Specifications
About this item
SG-Lysine-C™ endopeptidase, from Lysobacter enzymogenes, is a serine protease highly specific in cleaving peptide bonds at the carboxy side of lysine. Highly purified preparations of SG-Lysine-C™ are chemically modified making the enzyme resistant to autolysis and stabilizing its enzymatic activity.
SG-Lysine-C™ is a modified Lysine-C with high specificity, free from other known endopeptidases, that is resistant to autolysis and degradation
SG-Lysine-C™ is supplied lyophilized in 5 μg vials. The enzyme is typically reconstituted to a concentration of 0.25 μg/mL. For fragmentation, the enzyme is added to the sample protein in a ratio of 1:100 to 1:20 (enzyme to protein, by weight) in a standard digestion buffer.
SG-Chymotrypsin™ is used for the digestion of proteins for sequence analysis and is suitable for sequencing applications. Use of SG-Lysine-C™ produces consistent activity levels from lot to lot for reproducible digestions.