Diatomaceous earth, flux-calcined, Sigma-Aldrich®
Supplier: Sigma Aldrich
Certificates
About this item
CAS Number:61790-53-2
:7631-86-9
Formula:SiO₂
MDL Number:MFCD00132803
Boiling Pt:2200 °C (1013 hPa)
Melting Pt:1710 °C
Density:2.3 g/cm³ (20 °C)
:60.08 g/mol
:GHS08
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Appearance (Color):White (may contain low levels of black particles)
- Appearance (Form):Powder
- Particle Size Distribution:≤12.0 % (150 mesh Tyler sieve retain)
- Wet Density (lbs/ft3):≤ 21.5
Specification Test Results
Frequently Bought Together