Order Entry
United States
ContactUsLinkComponent
 
1,2,3-Trichloropropane, Standard, 2000 μg/ml, Restek
  RK30429
 :  Restek
 :  
undefined
1,2,3-Trichloropropane, Standard, 2000 μg/ml, Restek
  RK30429
 :  Restek
 :  30429
 :  

 

  • Pack type:
    Ampule
  • CAS No.:
    96-18-4
  • Size:
    5×1 ml
  • Formula:
    C₃H₅Cl₃
  • Description:
    1,2,3-Trichloropropane
  • Cat. No.:
    RK30429
  • Supplier no.:
    30429

 

 

Solvent packaged in purge-and-trap-grade methanol.

  • 2000 μg/mL in P&T methanol, 1 mL/ampoule

Packaged in purge-and-trap-grade methanol at 2000 μg/mL. Each standard includes an ampoule breaker, deactivated amber screw-top vial, extra product label, Certificate of Analysis and Material Safety Data Sheet.