Chloromethane (Methyl chloride), Standard, 2000 μg/ml, Restek
Catalog # RK30267
Supplier: Restek
CAS Number:
Specifications
- Pack type:Ampule
- CAS No.:74-87-3
- Size:1 ml
- Formula:CH₃Cl
- Description:Chloromethane (Methyl Chloride)
- Cat. No.:RK30267
- Supplier no.:30267
Specifications
About this item
Solvent packaged in purge-and-trap-grade methanol.
- 2000 μg/mL in P&T methanol, 1 mL/ampoule
Packaged in purge-and-trap-grade methanol at 2000 μg/mL. Each standard includes an ampoule breaker, deactivated amber screw-top vial, extra product label, Certificate of Analysis and Material Safety Data Sheet.