Bromoform, Standard, 2000 μg/ml, Restek
RK30252
: Restek
:
- Pack type:Ampule
- CAS No.:75-25-2
- Size:1 ml
- Formula:CHBr₃
- Description:Bromoform
- Cat. No.:RK30252
- Supplier no.:30252
Solvent packaged in purge-and-trap-grade methanol.
- 2000 μg/mL in P&T methanol, 1 mL/ampoule
Packaged in purge-and-trap-grade methanol at 2000 μg/mL. Each standard includes an ampoule breaker, deactivated amber screw-top vial, extra product label, Certificate of Analysis and Material Safety Data Sheet.